Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MSIRVTQKS YKVSTSGPRA FSSRSYTSGP GSRISSSSFS RVGSSNFRGG LGGGYGGASG MGGITAVTVN QSLLSPLVLE VDPNIQAVRT QEKEQIKTLN NKFASFIDKV RFLEQQNKML ETKWSLLQQQ KTARSNMDNM FESYINNLRR QLETLGQEKL KLEAELGNMQ GLVEDFKNKY EDEINKRTEM ENEFVLIKKD VDEAYMNKVE LESRLEGLTD EINFLRQLYE EEIRELQSQI SDTSVVLSMD NSRSLDMDSI IAEVKAQYED IANRSRAEAE SMYQIKYEEL QSLAGKHGDD LRRTKTEISE MNRNISRLQA EIEGLKGQRA SLEAAIADAE QRGELAIKDA NAKLSELEAA LQRAKQDMAR QLREYQELMN VKLALDIEIA TYRKLLEGEE SRLESGMQNM SIHTKTTSGY AGGLSSAYGG LTSPGLSYSL GSSFGSGAGS SSFSRTSSSR AVVVKKIETR DGKLVSESSD VLPK (1-483 aa encoded by BC000654)
Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-8; cytokeratin 8; Cytokeratin endo A; Cytokeratin-8; K8; keratin 8, type II; keratin complex 2, basic, gene 8; Keratin, type II cytoskeletal 8; Keratin-8; Type-II keratin Kb8
Gene Aliases: AA960620; AL022697; AU019895; CARD2; CK-8; CK8; CYK8; CYKER; EndoA; K2C8; K8; KO; Krt-2.8; Krt2-8; KRT8
UniProt ID: (Human) P05787, (Mouse) P11679, (Rat) Q10758
Entrez Gene ID: (Human) 3856, (Mouse) 16691, (Rat) 25626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support