Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLPRRPLAW PAWLLRGAPG AAGSWGRPVG PLARRGCCSA PGTPEVPLTR ERYPVQRLPF STVSKQDLAA FERIVPGGVV TDPEALQAPN VDWLRTLRGC SKVLLRPRTS EEVSHILRHC HERNLAVNPQ GGNTGMVGGS VPVFDEIILS TARMNRVLSF HSVSGILVCQ AGCVLEELSR YVEERDFIMP LDLGAKGSCH IGGNVATNAG GLRFLRYGSL HGTVLGLEVV LADGTVLDCL TSLRKDNTGY DLKQLFIGSE GTLGIITTVS ILCPPKPRAV NVAFLGC (1-286 aa encoded by BC036604)
This gene encodes D-2hydroxyglutarate dehydrogenase, a mitochondrial enzyme belonging to the FAD-binding oxidoreductase/transferase type 4 family. This enzyme, which is most active in liver and kidney but also active in heart and brain, converts D-2-hydroxyglutarate to 2-ketoglutarate. Mutations in this gene are present in D-2-hydroxyglutaric aciduria, a rare recessive neurometabolic disorder causing developmental delay, epilepsy, hypotonia, and dysmorphic features.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-2-hydroxyglutarate dehydrogenase, mitochondrial; FLJ42195; MGC25181
Gene Aliases: AA408776; AA408778; AI325464; D2HGD; D2HGDH
UniProt ID: (Human) Q8N5Q8, (Mouse) Q8CIM3
Entrez Gene ID: (Human) 728294, (Mouse) 98314
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support