Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: KSLSMVDLQD ARTLDGEAGS PAGPDVLPTD GQAAAAQLVA GWPARATPVN LAGLATVRRA GQTPTTPGTS EGAPGRPQLL APLSFQNPVY QMAAGLPLSP
DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIP-1; ASK-interacting protein 1; ASK1-interacting protein 1; DAB2 interaction protein; Disabled homolog 2-interacting protein; DOC-2/DAB-2 interactive protein; DOC-2/DAB2 interactive protein; nGAP-like protein
Gene Aliases: AF9Q34; AIP-1; AIP1; DAB2IP; DIP1/2; KIAA1743
UniProt ID: (Human) Q5VWQ8
Entrez Gene ID: (Human) 153090
Molecular Function: GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support