Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This antibody is predicted to react with rat based on sequence homology.
This gene encodes a diacylglycerol lipase. The encoded enzyme is involved in the biosynthesis of the endocannabinoid 2-arachidonoyl-glycerol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAGL-alpha; DGL-alpha; Diacylglycerol lipase-alpha; Neural stem cell-derived dendrite regulator; Sn1-specific diacylglycerol lipase alpha
Gene Aliases: C11orf11; DAGL(ALPHA); DAGLA; DAGLALPHA; KIAA0659; NSDDR
UniProt ID: (Human) Q9Y4D2, (Mouse) Q6WQJ1
Entrez Gene ID: (Human) 747, (Mouse) 269060
Molecular Function:
hydrolase
lipase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support