Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: VRRIKMATA DEIVKLMLDH MTNTTNASHV PVQPGSSVVM MVNNLGGLSF LELGIIADAT VRSLEGRGVK IARALVGTFM SALEMPGISL TLLLVDEPLL KLIDAETTAA AWPNVAAVSI TGRKRSRVAP AEPQEAPDST AAGGSASKRM ALVLERVCST LLGLEEHLNA LDRAAGDGDC GTTHSRAARA IQEWLKEGPP PASPAQLLSK LSVLLLEKMG GSSGALYGLF LTAAAQPLKA KTSLPAWSAA MDAGLEAMQK YGKAAPGDRT MLDSLWAAGQ ELQAWKSPGA DLLQVLTKAV KSAEAAAEAT KNMEAGAGRA SYISSARLEQ PDPGAVAAAA ILRAILEVLQ S (226-576 aa encoded by BC001341)
This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4',5'-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent dihydroxyacetone kinase; Bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing); DHA kinase; Dha kinase/FMN cyclase; dihydroxyacetone kinase 2 homolog; DKFZP586B1621; FAD-AMP lyase cyclic FMN forming; FAD-AMP lyase cyclizing; glycerone kinase; MGC5621; testis tissue sperm-binding protein Li 84P; Triokinase/FMN cyclase
Gene Aliases: BC021917; DAK; NET45; RGD1311026; TKFC
UniProt ID: (Rat) Q4KLZ6, (Human) Q3LXA3, (Mouse) Q8VC30
Entrez Gene ID: (Rat) 361730, (Human) 26007, (Mouse) 225913
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support