Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTSKKLVNSV AGCADDALAG LVACNPNLQL LQGHRVALRS DLDSLKGRVA LLSGGGSGHE PAHAGFIGKG MLTGVIAGAV FTSPAVGSIL AAIRAVAQAG TVGTLLIVKN YTGDRLNFGL AREQARAEGI PVEMVVIGDD SAFTVLKKAG RRGLCGTVLI HKVAGALAEA GVGLEEIAKQ VNVVAKAMGT LGVSLSSCSV PGSKPTFELS ADEVELGLGI HGEAGVRRIK MATADEIVKL MLDHMTNTTN ASHVPVQPGS SVVMMVNNLG GLSFLELGII ADATVRSLEG RGVKIARALV GTFMSALEMP GISLTLLLVD EPLLKLIDAE TTAAAWPNVA AVSITGRKRS RVAPAEPQEA PDSTAAGGSA SKRMALVLER VCSTLLGLEE HLNALDRAAG DGDCGTTHSR AARAIQEWLK EGPPPASPAQ LLSKLSVLLL EKMGGSSGAL YGLFLTAAAQ PLKAKTSLPA WSAAMDAGLE AMQKYGKAAP GDRTMLDSLW AAGQELQAWK SPGADLLQVL TKAVKSAEAA AEATKNMEAG AGRASYISSA RLEQPDPGAV AAAAILRAIL EVLQS
This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4',5'-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent dihydroxyacetone kinase; Bifunctional ATP-dependent dihydroxyacetone kinase/FAD-AMP lyase (cyclizing); DHA kinase; Dha kinase/FMN cyclase; dihydroxyacetone kinase 2 homolog; DKFZP586B1621; FAD-AMP lyase cyclic FMN forming; FAD-AMP lyase cyclizing; glycerone kinase; MGC5621; testis tissue sperm-binding protein Li 84P; Triokinase/FMN cyclase
Gene Aliases: DAK; NET45; TKFC
UniProt ID: (Human) Q3LXA3
Entrez Gene ID: (Human) 26007
Molecular Function:
cyclase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support