Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PRELDMFPDY GYGWFHTSLI LEGKNYLQWL TERLTERGVK FFQRKVESFE EVAREGADVI VNCTGVWAGA LQRDPLLQPG RGQIMKVDAP WMKHFILTHD
This gene encodes the peroxisomal enzyme D-amino acid oxidase. The enzyme is a flavoprotein which uses flavin adenine dinucleotide (FAD) as its prosthetic group. Its substrates include a wide variety of D-amino acids, but it is inactive on the naturally occurring L-amino acids. Its biological function is not known; it may act as a detoxifying agent which removes D-amino acids that accumulate during aging. In mice, it degrades D-serine, a co-agonist of the NMDA receptor. This gene may play a role in the pathophysiology of schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABP; Abp1; AOC1; D-amino-acid oxidase; DAAO; Diamine oxidase; Histaminase; KAO
Gene Aliases: DAAO; DAMOX; DAO; OXDA
UniProt ID: (Human) P14920
Entrez Gene ID: (Human) 1610
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support