Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
DAPK1 is a large, calmodulin-dependent kinase involved in gamma-interferon induced programmed cell death and is a potential tumor suppressor. DAPK1 is involved in multiple cellular signaling pathways that trigger cell survival, apoptosis, and authophagy. It regulates both type I apoptotic and type II autophagic cell death signal, depending on the cellular setting. Type I apoptotic cell death is caspase-dependent, type II authophagic cell death is caspase-independent. DAPK1 phosphorylates PIN1 resulting in inhibition of its catalytic activity, nuclear localization, and cellular function. It also phosphorylates TPM1, STX1A, PRKD1, BECN1, TSC2, RPS6, MYL9, and DAPK3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAP kinase 1; Death-associated protein kinase 1; DKFZp781I035; OTTHUMP00000216193
Gene Aliases: D13Ucla1; DAP-Kinase; DAPK; DAPK1
UniProt ID: (Human) P53355, (Mouse) Q80YE7
Entrez Gene ID: (Human) 1612, (Mouse) 69635
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support