Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
DAPK2 is a calcium/calmodulin-dependent kinase that acts as a positive regulator of apoptosis, and is expressed in many tissue types. DAPK2 encodes a protein that belongs to the serine/threonine protein kinase family. This protein contains a N-terminal protein kinase domain followed by a conserved calmodulin-binding domain with significant similarity to that of death-associated protein kinase 1 (DAPK1), a positive regulator of programmed cell death. Overexpression of this gene was shown to induce cell apoptosis. It uses multiple polyadenylation sites.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAP kinase 2; DAP-kinase-related protein 1; DAP-kinase-related protein 1 beta isoform; death-associated kinase 2; Death-associated protein kinase 2; DRAK2; DRP-1; MGC119312
Gene Aliases: DAPK2; DRP-1; DRP1
UniProt ID: (Human) Q9UIK4, (Mouse) Q8VDF3
Entrez Gene ID: (Human) 23604, (Mouse) 13143
Molecular Function: non-receptor serine/threonine protein kinase immunoglobulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support