Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QLDDAVRPE AEGEEEGRAT VNQDTRLDNR VIDLRTSTSQ AVFRLQSGIC HLFRETLINK GFVEIQTPKI ISAASEGGAN VFTVSYFKNN AYLAQSPQLY KQMCICADFE KVFSIGPVFR AEDSNTHRHL TEFVGLDIEM AFNYHYHEVM EEIADTMVQI FKGLQERFQT EIQTVNKQFP CEPFKFLEPT LRLEYCEALA MLREAGVEMG DEDDLSTPNE KLLGHLVKEK YDTDFYILDK YPLAVRPFYT MPDPRNPKQS NSYDMFMRGE EILSGAQRIH DPQLLTERAL HHGIDLEKIK AYIDSFRFGA PPHAGGGIGL ERVTMLFLGL HNVRQTSMFP RDPKRLTP (155-501 aa encoded by BC000629)
Aspartyl-tRNA synthetase (DARS) is part of a multienzyme complex of aminoacyl-tRNA synthetases. Aspartyl-tRNA synthetase charges its cognate tRNA with aspartate during protein biosynthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aspartate tRNA ligase 1, cytoplasmic; Aspartate--tRNA ligase, cytoplasmic; Aspartyl-tRNA synthetase; aspartyl-tRNA synthetase, cytoplasmic; AspRS; Cell proliferation-inducing gene 40 protein; testicular tissue protein Li 192
Gene Aliases: 5730439G15Rik; aspRS; DARS; DARS1; DRS1; HBSL; PIG40
UniProt ID: (Human) P14868, (Rat) P15178, (Mouse) Q922B2
Entrez Gene ID: (Human) 1615, (Rat) 116483, (Mouse) 226414
Molecular Function: aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support