Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Immunogen sequence: MSTANPETP NSTISREAST QSSSAATSQG YILPEGKIMP NTVFVGGIDV RMDETEIRSF FARYGSVKEV KIITDRTGVS KGYGFVSFFN DVDVQKIVES QINFHGKKLK LGPAIRKQNL CAYHVQPRPL VFNHPPPPQF QNVWTNPNTE TYMQPTTTMN PITQYVQAYP TYPNSPVQVI TGYQLPVYNY QMPPQWPVGE QRSYVVPPAY SAVNYHCNEV DPGAEVVPNE CSVHEATPPS GNGPQKKSVD RSIQTVVSCL FNPENRLRNS VVTQDDYFKD KRVHHFRRSR AMLKSV (1-295 aa encoded by B C027595)
Protein expression pattern show that DAZL is involved in mitosis and meiosis of human germ cell. The DAZL gene family encodes potential RNA binding proteins that are expressed in prenatal and postnatal germ cells of males and females.The protein is expressed in germ cells of diverse mammalian species. The protein encoded by this gene is localized to the nucleus and cytoplasm of fetal germ cells and to the cytoplasm of developing oocytes. In humans, DAZL is expressed in primordial germ cells, adult testis, and the ovary. It plays a central role during spermatogenesis. Defects in DAZL may cause infertility due to severe oligozoospermia (az) and non obstructive azoospermia. The concentrations of DAZL transcripts were lower in men with spermatogenic failure. Drosophila and mouse mutants that have lost DAZ homologous gene function found to be azoospermic, just as with human azoospermia DAZ. Transposition and amplification of this autosomal gene during primate evolution gave rise to the DAZ gene cluster on the Y chromosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAZ homolog; DAZ-like autosomal; deleted in azoospermia protein 3; Deleted in azoospermia-like; Deleted in azoospermia-like 1; germline specific RNA binding protein; HGNC:2685; MGC26406; spermatogenesis gene on the Y-like autosomal; SPGY-like-autosomal; testis secretory sperm-binding protein Li 204a
Gene Aliases: Daz-like; DAZH; DAZL; DAZL1; DAZLA; SPGYLA; Tpx-2; Tpx2
UniProt ID: (Human) Q92904, (Mouse) Q64368
Entrez Gene ID: (Human) 1618, (Mouse) 13164, (Rat) 680486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support