Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
A suggested positive control is 293 cell lysate.
PA5-20939 can be used with blocking peptide PEP-1053.
Cancer-testis (CT) antigens are a type of antigen whose expression is highly restricted, primarily in normal testis, but aberrantly in different tumor types. Their immunogenicity in cancer patients makes them prime targets for immunotherapy of human tumors. One such CT antigen is TCC52, a novel centrosomal protein. TCC52 is a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. The normal biological function of this protein is not known at this time.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 102; Centrosome-related protein TCC52; DDB1- and CUL4-associated factor 12; similar to WD repeat domain 40A; Testis cancer centrosome-related protein; WD repeat domain 40A; WD repeat-containing protein 40A
Gene Aliases: 1500001L20Rik; 5830424K06Rik; AA420338; AI851081; CT102; DCAF12; KIAA1892; TCC52; WDR40A
UniProt ID: (Human) Q5T6F0, (Mouse) Q8BGZ3
Entrez Gene ID: (Human) 25853, (Mouse) 68970, (Rat) 682812
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support