Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: DIPLLFETKK LLKYMKHTVV VYCDRDTQLA RLMRRNSLNR KDAEARINAQ LPLTDKARMA RHVLDNSGEW SVTKRQVILL HTELERSL
DCAKD is a protein coding gene. Gene ontology (GO) annotation include membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dephospho-CoA kinase domain-containing protein
Gene Aliases: DCAKD
UniProt ID: (Human) Q8WVC6
Entrez Gene ID: (Human) 79877
Molecular Function: kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support