Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATPPKRSC PSFSASSEGT RIKKISIEGN IAAGKSTFVN ILKQLCEDWE VVPEPVARWC NVQSTQDEFE ELTMSQKNGG NVLQMMYEKP ERWSFTFQTY ACLSRIRAQL ASLNGKLKDA EKPVLFFERS VYSDRYIFAS NLYESECMNE TEWTIYQDWH DWMNNQFGQS LELDGIIYLQ ATPETCLHRI YLRGRNEEQG IPLEYLEKLH YKHESWLLHR TLKTNFDYLQ EVPILTLDVN EDFKDKYESL VEKVKEFLST L (1-260 aa encoded by BC103764)
Deoxycytidine kinase (DCK) is required for the phosphorylation of several deoxyribonucleosides and their nucleoside analogs. Deficiency of DCK is associated with resistance to antiviral and anticancer chemotherapeutic agents. Conversely, increased deoxycytidine kinase activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. DCK is clinically important because of its relationship to drug resistance and sensitivity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dCK; Deoxyadenosine kinase; Deoxycytidine kinase; Deoxyguanosine kinase; deoxynucleoside kinase
Gene Aliases: DCK
UniProt ID: (Human) P27707
Entrez Gene ID: (Human) 1633
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support