Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 11
Immunogen sequence: QGVVRSLSY EEPRRHSPPI EKQLCPAIQK LMVRSADLHP LSELPENRPC ENGSTHSAGE FFTGPVQPGS PHNIGTSRGV QNASRTQNLF EKLQSTPGAA NKCDPSTPAP ASSAALNRSR APTSVTPVAP GKGLAQPPQA YFNGSLPPQT VGHQAHGREQ STLPRQTLAI SGSQTGSSGV ISPQELLKKL QIVQQEQQLH ASNRPALAAK FPVLAQSSGT GKPLESWINK TPNTEQQTPL FQVISPQRIP ATAAPSLLMS PMVFAQPTSV PPKERESGLL PVGGQEPPAA ATSLLLPIQS PEPSVITSSP LTKLQLQEAL LYLIQNDDNF LNIIYEAYLF SMTQAAMKKT M (269-618 aa encoded by BC043437)
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCP1 decapping enzyme homolog B; decapping enzyme hDcp1b; decapping mRNA 1B; hDcp1b; mRNA-decapping enzyme 1B
Gene Aliases: B930050E02Rik; DCP1; DCP1B; RGD1562214
UniProt ID: (Human) Q8IZD4
Entrez Gene ID: (Human) 196513, (Rat) 500305, (Mouse) 319618
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support