Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
DCP1B is a core component of the mRNA decapping complex, a key factor in the regulation of mRNA decay.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCP1 decapping enzyme homolog B; decapping enzyme hDcp1b; hDcp1b; mRNA-decapping enzyme 1B
Gene Aliases: DCP1; DCP1B
UniProt ID: (Human) Q8IZD4
Entrez Gene ID: (Human) 196513
Molecular Function:
mRNA capping factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support