Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MNASEFRRRG KEMVDYVANY MEGIEGRQVY PDVEPGYLRP LIPAAAPQEP DTFEDIINDV EKIIMPGVTH WHSPYFFAYF PTASSYPAML ADMLCGAIGC IGFSWAASPA CTELETVMMD WLGKMLELPK AFLNEKAGEG GGVIQGSASE ATLVALLAAR TKVIHRLQAA SPELTQAAIM EKLVAYSSDQ AHSSVERAGL IGGVKLKAIP SDGNFAMRAS ALQEALERDK AAGLIPFFMV ATLGTTTCCS FDNLLEVGPI CNKEDIWLHV DAAYAGSAFI CPEFRHLLNG VEFADSFNFN PHKWLLVNFD CSAMWVKKRT DLTGAFRLDP TYLKHSHQDS GLITDYRHWQ IPLGRRFRSL KMWFVFRMYG VKGLQAYIRK HVQLSHEFES LVRQDPRFEI CVEVILGLVC FRLKGSNKVN EALLQRINSA KKIHLVPCHL RDKFVLRFAI CSRTVESAHV QRAWEHIKEL AADVLRAERE
DOPA Decarboxylase (aromatic L-amino acid decarboxylase, AADC; DDC) catalyzes the second reaction in the biosynthesis of catecholamines and serotonin (Waymire and Haycock, 2002; Berry et al., 1996; Haycock et al., 2003). It is also involved in the biosynthesis of trace amines. DDC antibodies can therefore be used as markers for dopaminergic, noradrenergic and serotonergic neurons in a variety of applications including depression, schizophrenia, Parkinson's disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AADC; aromatic L-amino acid decarboxylase; Aromatic-L-amino-acid decarboxylase; DDC; DOPA decarboxylase; dopa decarboxylase (aromatic L-amino acid decarboxylase); HGNC:2719
Gene Aliases: AADC; DDC
UniProt ID: (Human) P20711
Entrez Gene ID: (Human) 1644
Molecular Function:
decarboxylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support