Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognizes DDIT4L / REDD2 protein, which belongs to intracellular stress-induced proteins involved in mediation of cell death.
DDIT4L inhibits cell growth by regulating the TOR signaling pathway upstream of the TSC1-TSC2 complex and downstream of AKT1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like; HIF-1 responsive protein RTP801-like; homolog of mouse SMHS1; Protein regulated in development and DNA damage response 2; REDD-2; regulated in development and DNA damage response 2
Gene Aliases: DDIT4L; REDD2; RTP801L
UniProt ID: (Human) Q96D03
Entrez Gene ID: (Human) 115265
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support