Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRPARHWET RFGARDFGGF QDCFFRDRLM DTARIAVVGA GVVGLSTAVC ISKLVPRCSV TIISDKFTPD TTSDVAAGML IPHTYPDTPI HTQKQWFRET FNHLFAIANS AEAGDAGVHL VSGWQIFQST PTEEVPFWAD VVLGFRKMTE AELKKFPQYV FGQAFTTLKC ECPAYLPWLE KRIKGSGGWT LTRRIEDLWE LHPSFDIVVN CSGLGSRQLA GDSKIFPVRG QVLQVQAPWV EHFIRDGSGL TYIYPGTSHV TLGGTRQKGD WNLSPDAENS REILSRCCAL EPSLHGACNI REKVGLRPYR PGVRLQTELL ARDGQRLPVV HHYGHGSGGI SVHWGTALEA ARLVSECVHA LRTPIPKSNL (1-369 aa encoded by BC032786)
DDO is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aspartic oxidase; D-aspartate oxidase; D-aspartate oxidase, DDO; DASOX
Gene Aliases: 5330420D20Rik; 5730402C02Rik; AI467244; DASOX; DDO; DDO-1; DDO-2; LOC100911156
UniProt ID: (Human) Q99489, (Rat) D3ZDM7, (Mouse) Q922Z0
Entrez Gene ID: (Human) 8528, (Rat) 685325, (Mouse) 70503
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support