Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-dopachrome decarboxylase; D-dopachrome tautomerase; hCG_41098; Phenylpyruvate tautomerase II
Gene Aliases: DDCT; DDT
UniProt ID: (Human) P30046
Entrez Gene ID: (Human) 1652
Molecular Function: decarboxylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support