Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SHSECGIIE KATSPKELGC DRQSEEQMKN SVQTPVENST NSQHQVKEAL PVSLPQIPCL SSFKIHQPYT LTFAELVEDY EHYIKEGLEK PVEIIRHYTG PGDQTVNPQN GFVRNKVTEQ RVPVLASSSQ SGDSESDSDS YSSRTSSQSK GNKSYLEGSS DNQLKDSEST PVDDRISLEQ PPNGSDTPNP EKYQESPGIQ MKTRLKEGAS QRAKQSRRNL PRRSSFRLQT EAQEDDWYDC HREIRLSFSD TYQDYEEYWR AYYRAWQEYY AAASHSYYWN AQRHPSWMAA YHMNTIYLQE MMHSNQ (520-824 aa encoded by BC011556)
Gemin 3 is a member of the DEAD box protein family which are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. DEAD box proteins such as Gemin 3 are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of DEAD box family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Gemin 3 has an ATPase activity and a component of the survival of motor neurons (SMN) complex. The Gemin 3 protein may play a catalytic role in the function of the SMN complex on RNPs. Diseases associated with Gemin 3 protein dysfunction include spinal muscular dystrophy and muscular atrophy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Component of gems 3; DDX 20; DEAD (Asp-Glu-Ala-Asp) box polypeptide 20; DEAD box protein 20; DEAD box protein DP 103; DEAD-box protein DP103; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 20, 103kD; DEAD/H BOX 20; DKFZP434H052; DP 103; Gemin-3; Probable ATP-dependent RNA helicase DDX20; Regulator of steroidogenic factor 1; ROSF-1; SMN-interacting protein
Gene Aliases: DDX20; DP103; GEMIN3
UniProt ID: (Human) Q9UHI6, (Mouse) Q9JJY4
Entrez Gene ID: (Human) 11218, (Mouse) 53975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support