Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAD (Asp-Glu-Ala-Asp) box helicase 21; DEAD (Asp-Glu-Ala-Asp) box polypeptide 21; DEAD box protein 21; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 21; Gu protein; Gu-alpha; Nucleolar RNA helicase 2; Nucleolar RNA helicase Gu; Nucleolar RNA helicase II; RH II/Gu; RNA helicase II/Gu alpha
Gene Aliases: DDX21; GUA; GURDB; RH-II/GU; RH-II/GuA
UniProt ID: (Human) Q9NR30
Entrez Gene ID: (Human) 9188
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support