Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: ELRHLLSQPL FTESQKTKYP TQSGKPPLLV SAPSKSESAL SCLSKQKKKK
Sequence homology: Human: 100%; Mouse: 79%; Rat: 79%
METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; S. cerevisiae CHL1-like helicase
Gene Aliases: DDX24
UniProt ID: (Human) Q9GZR7
Entrez Gene ID: (Human) 57062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support