Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
ATP-binding RNA helicase involved in the biogenesis of 60S ribosomal subunits. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DDX51; DEAD (Asp-Glu-Ala-Asp) box polypeptide 51; DEAD box protein 51
Gene Aliases: DDX51
UniProt ID: (Human) Q8N8A6
Entrez Gene ID: (Human) 317781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support