Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LRKYNDALI ISEHARMKDA LDYLKDFFSN VRAAGFDEIE QDLTQRFEEK LQELESVSRD PSNENPKLED LCFILQEEYH LNPETITILF VKTRALVDAL KNWIEGNPKL SFLKPGILTG RGKTNQNTGM TLPAQKCILD AFKASGDHNI LIATSVADEG IDIAQCNLVI LYEYVGNVIK MIQTRGRGRA RGSKCFLLTS NAGVIEKEQI NMYKEKMMND SILRLQTWDE AVFREKILHI QTHEKFIRDS QEKPKPVPDK ENKKLLCRKC KALACYTADV RVIEECHYTV LGDAFKECFV SRPHPKPKQF SSFEKRAKIF CARQNCSHDW GIHVKYKTFE IPVIKIESFV VEDIATGVQT LYSKWKDFHF EKIPFDPAEM SK (545-925 aa encoded by BC132786)
Retinoic acid-inducible gene I, RIG-I is a pattern recognition receptor (PRR) involved in the recognition of viral dsRNA. Along with MDA5, RIG-I detects viral dsRNA and activates the innate immune response. Both MDA5 and RIG-I are RNA helicases and they perform overlapping as well as distinct roles. RIG-I is activated by dsRNAs without a 5'-triphosphate end and short dsRNAs, whereas MDA5 is activated by long dsRNAs. Once activated, both proteins signal through IPS-1 activating transcription factors NF-kappaB and IRF-3 (1) and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus (3), paromyxoviruses, Japanese encephalitis virus, and West Nile virus. MicroRNA-146a has been implicated in feedback inhibition of RIG-I-dependant antiviral response by negatively regulating RIG-I targets TRAF6, IRAK1, and IRAK2. Recent evidence has implicated RIG-I in the detection of cytosolic DNA through RNA polymerase III activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antiviral innate immune response receptor RIG-I; ATP-dependent RNA helicase DDX58; DEAD (Asp-Glu-Ala-Asp) box polypeptide 58; DEAD box protein 58; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; Retinoic acid-inducible gene 1 protein; Retinoic acid-inducible gene I protein; RIG-1; RIG-I; RIG-I-like receptor 1; RLR-1; RNA helicase RIG-I; RNA sensor RIG-I
Gene Aliases: DDX58; RIG-I; RIGI; RLR-1; SGMRT2
UniProt ID: (Human) Q9NT04
Entrez Gene ID: (Human) 23586
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support