Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSTARTENPV IMGLSSQNGQ LRGPVKPTGG PGGGGTQTQQ QMNQLKNTNT INNGTQQQAQ SMTTTIKPGD DWKKTLKLPP KDLRIKTSDV TSTKGNEFED YCLKRELLMG IFEMGWEKPS PIQEESIPIA LSGRDILARA KNGTGKSGAY LIPLLERLDL KKDNIQAMVI VPTRELALQV SQICIQVSKH MGGAKVMATT GGTNLRDDIM RLDDTVHVVI ATPGRILDLI KKGVAKVDHV QMIVLDEADK LLSQDFVQIM EDIILTLPKN RQILLYSATF PLSVQKFMNS HLQKPYEINL MEELTLKGVT QYYAYVTERQ KVHCLNTLFS RLQINQSIIF CNSSQRVELL AKKISQLGYS CFYIHAKMRQ EHRNRVFHDF RNGLCRNLVC TDLFTRGIDI QAVNVVINFD FPKLAETYLH RIGRSGRFGH LGLAINLITY DDRFNLKSIE EQLGTEIKPI PSNIDKSLYV AEYHSEPVED EKP
This gene encodes a member of the DEAD box protein family. The protein is an RNA helicase found in P-bodies and stress granules, and functions in translation suppression and mRNA degradation. It is required for microRNA-induced gene silencing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase p54; DEAD (Asp-Glu-Ala-Asp) box helicase 6; DEAD (Asp-Glu-Ala-Asp) box polypeptide 6; DEAD box protein 6; DEAD box-6; DEAD-box protein 6; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 6 (RNA helicase, 54kD); Oncogene RCK; Probable ATP-dependent RNA helicase DDX6
Gene Aliases: DDX6; HLR2; P54; RCK
UniProt ID: (Human) P26196
Entrez Gene ID: (Human) 1656
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support