Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEDSDSAAK QLGLAEAAAV AAAAAVAAAA AAAAGGEAEE PVLSRDEDSE EDADSEAERE TPRVTAVAVM AAEPGHMDMG AEALPGPDEA AAAAAFAEVT TVTVANVGAA ADNVFTTSVA NAASISGHVL SGRTALQIGD SLNTEKATLI VVHTDGSIVE TTGLKGPAAP LTPGPQSPPT PLAPGQEKGG TKYNWDPSVY DSELPVRCRN ISGTLYKNRL GSGGRGRCIK QGENWYSPTE FEAMAGRASS KDWKRSIRYA GRPLQCLIQD GILNPHAASC TCAACCDDMT LSGPVRLFVP YKRRKKENEL PTTPVKK (1-316 aa encoded by BC053322)
HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAF-1 related transcriptional regulator (NUDR); DEAF1 transcription factor; Deformed epidermal autoregulatory factor 1 homolog; Nuclear DEAF-1-related transcriptional regulator; NUDR; Suppressin; Zinc finger MYND domain-containing protein 5
Gene Aliases: AU042387; C230009B13Rik; DEAF1; MRD24; NUDR; SPN; ZMYND5
UniProt ID: (Human) O75398, (Mouse) Q3UJA4
Entrez Gene ID: (Human) 10522, (Mouse) 54006, (Rat) 83632
Molecular Function: zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support