Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: Mouse spleen
Immunogen sequence: MRTLAILAAI LLVALQAQAE PLQARADEVA AAPEQIAADI PEVVVSLAWD ESLAPKHPGS RKNMACYCRI PACIAGERRY GTCIYQGRLW AFCC
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The protein encoded by this gene, defensin, alpha 1, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8. This gene differs from defensin, alpha 3 by only one amino acid. This gene and the gene encoding defensin, alpha 3 are both subject to copy number variation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-defensin 1; Cryptdin-1; defensin related cryptdin peptide 1; Defensin, alpha 1; defensin, alpha 1, myeloid-related sequence; defensin, alpha 2; defensin-1; Defensin-related cryptdin peptide; HNP-1; HP-1; myeloid-related sequence; Neutrophil defensin 1
Gene Aliases: DEF1; DEFA1; DEFA1B; DEFA2; Defcr; Defcr1; HNP-1; HP-1; HP1; MRS
UniProt ID: (Human) P59665, (Mouse) P11477
Entrez Gene ID: (Human) 1667, (Mouse) 13216
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support