Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRIHYLLFAL LFLFLVPVPG HGGIINTLQK YYCRVRGGRC AVLSCLPKEE QIGKCSTRGR KCCRRKK
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BD-3; beta defensin 3; beta defensin-3; Beta-defensin 103; Beta-defensin 3; Defensin, beta 103; defensin, beta 103A; defensin, beta 3; Defensin-like protein
Gene Aliases: BD-3; BD3; DEFB-3; DEFB103; DEFB103A; DEFB103B; DEFB3; hBD-3; HBD3; HBP-3; HBP3
UniProt ID: (Human) P81534
Entrez Gene ID: (Human) 414325
Molecular Function: antimicrobial response protein defense/immunity protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support