Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This target displays homology in the following species: Dog: 77%; Guinea Pig: 82%; Horse: 92%; Human: 100%; Pig: 92%; Rat: 85%
This gene encodes a member of the beta subfamily of defensins. Beta-defensins are antimicrobial peptides that protect tissues and organs from infection by a variety of microorganisms. This gene is found in a cluster with other beta-defensin genes on the long arm of chromosome 20. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-defensin 119; Beta-defensin 120; Beta-defensin 19; Beta-defensin 20; DEFB-19; DEFB-20; Defensin, beta 119; Defensin, beta 120; defensin, beta 19; defensin, beta 20; ESC42-RELA
Gene Aliases: DEFB-19; DEFB-20; DEFB119; DEFB120; DEFB19; DEFB20; ESC42-RELA; ESC42-RELB; UNQ2449/PRO5729
UniProt ID: (Human) Q8N690
Entrez Gene ID: (Human) 245932
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support