Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MSASAPAAEG EGTPTQPASE KEPEMPGPRE ESEEEEDEDD EEEEEEEKEK SLIVEGKREK KKVERLTMQV SSLQREPFTI AQGKGQKLCE IERIHFFLSK KKTDELRNLH KLLYNRPGTV SSLKKNVGQF SGFPFEKGSV QYKKKEEMLK KFRNAMLKSI CEVLDLERSG VNSELVKRIL NFLMHPKPSG KPLPKSKKTC SKGSKKERNS SGMARKAKRT KCPEILSDES SSDEDEKKNK EESSDDEDKE SEEEPPKKTA KREKPKQKAT SKSKKSVKSA NVKKADSSTT KKNQNSSKKE SESEDSSDDE PLIKKLKKPP TDEELKETIK KLLASANLEE VTMKQICKKV YENYPTYDLT ERKDFIKTTV KELIS
DEK is a protein with one SAP domain. This protein binds to cruciform and superhelical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEK oncogene (DNA binding); Protein DEK
Gene Aliases: 1810019E15Rik; D13H6S231E; D6S231E; DEK
UniProt ID: (Human) P35659, (Rat) Q6AXS3, (Mouse) Q8BZV6
Entrez Gene ID: (Human) 7913, (Rat) 306817, (Mouse) 110052
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support