Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: PSSLEDPRAR PPKALLAERA HLQPREEPGA LNSPATPTSN CQKSQPSSRP
Sequence homology: Human: 100%
Clathrin-mediated endocytosis is a major mechanism for internalization of proteins and lipids. Members of the connecdenn family, such as DENND1C, function as guanine nucleotide exchange factors (GEFs) for the early endosomal small GTPase RAB35 and bind to clathrin and clathrin adaptor protein-2. Thus, connecdenns link RAB35 activation with the clathrin machinery.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Connecdenn 3; DENN domain-containing protein 1C; DENN/MADD domain containing 1C; family with sequence similarity 31, member C; Protein FAM31C
Gene Aliases: DENND1C; FAM31C
UniProt ID: (Human) Q9H5Z2
Entrez Gene ID: (Human) 79958
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support