Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Bifunctional enzyme involved in flavonoid metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DFR; Dihydroflavonol 4-reductase; Dihydrokaempferol 4-reductase; Flavanone 4-reductase; FNR; Protein TRANSPARENT TESTA 3; TT3
Gene Aliases: AT5G42800; DFR; DFRA; DIHYDROFLAVONOL 4-REDUCTASE; DIHYDROKAEMPFEROL 4-REDUCTASE; M318; MJB21.18; MJB21_18; TT3
UniProt ID: (Arabidopsis) P51102
Entrez Gene ID: (Arabidopsis) 834291
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support