Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Diacylglycerol (DAG) is an allosteric activator of protein kinase C. DAG also participates in regulating RAS and RHO family proteins by activating the guanine nucleotide exchange factors VAV and RASGRP1. DAG is also involved in the synthesis of phospholipids and triacylglycerols. Tight regulation of DAG levels is achieved via DAG kinases (DGKs), which remove DAG by phosphorylate it to phosphatidic acid. Several mammalian isozymes of DAGK have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90 kDa diacylglycerol kinase; DAG kinase beta; DGK-beta; Diacylglycerol kinase beta; diacylglycerol kinase, beta 90kDa; Diglyceride kinase beta
Gene Aliases: 6430574F24; 90kda; C630029D13Rik; DAGK2; DGK; DGK-BETA; DGKB; KIAA0718; mKIAA0718
UniProt ID: (Human) Q9Y6T7, (Mouse) Q6NS52
Entrez Gene ID: (Human) 1607, (Mouse) 217480
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support