Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: CKQQCGCQPK LCDYRCIWCQ KTVHDECMKN SLKNEKCDFG EFKNLIIPPS YLTSINQMRK DKKTDYEVLA SKLGKQWTPL IILANSRSGT NMGEGLLGEF
Diacylglycerol (DAG) influences numerous cell signaling cascades by functioning as an intracellular, allosteric activator of protein kinase C (PKC), and as a potent activator of guanine nucleotide exchange factors. In order to maintain cellular homeostasis, intracellular DAG levels are tightly regulated by diacylglycerol kinases (DGKs, DAGKs), which phosphorylate DAG to phosphatidic acid, thus removing DAG. Human DGK-alpha (80 kDa), -beta (90 kDa), and - gamma (90 kDa) have calcium-binding EF-hand motifs at their N termini and are classified as type I DGKs. Human DGK- delta (130 kDa) and DGK-i (130 kDa) contain N-terminal pleckstrin homology (PH) domains and are classified as type II. Human DGK- epsilon (64 kDa) contains no identifiable regulatory domains and is classified as a type III DGK. Human DGK- zeta (104 kDa) and -iota (130 kDa) possess C-terminal ankyrin repeats and are classified as type IV DGKs. Human DGK-theta (110 kDa) contains 3 cysteine-rich domains and a PH domain and is classified as a type V DGK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAG kinase epsilon; DGK-epsilon; Diacylglycerol kinase epsilon; diacylglycerol kinase, epsilon 64kDa; Diglyceride kinase epsilon; EC 2.7.1.107
Gene Aliases: AHUS7; C87606; DAGK5; DAGK6; DGK; DGKE; NPHS7
UniProt ID: (Human) P52429, (Mouse) Q9R1C6
Entrez Gene ID: (Human) 8526, (Mouse) 56077, (Rat) 497978
Molecular Function: kinase transferase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support