Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAPMNGQV CVVTGASRGI GRGIALQLCK AGATVYITGR HLDTLRVVAQ EAQSLGGQCV PVVCDSSQES EVRSLFEQVD REQQGRLDVL VNNAYAGVQT ILNTRNKAFW ETPASMWDDI NNVGLRGHYF CSVYGARLMV PAGQGLIVVI SSPGSLQYMF NVLYGVGKAA CDKLAADCAH ELRRHGVSCV SLWPGIVQTE LLKEHMAKEE VLQDPVLKQF KSAFSSAETT ELSGKCVVAL ATDPNILSLS GKVLPSCDLA RRYGLRDVDG RPVQDYLSLS SVLSHVSGLG WLASYLPSFL RVPKWIIALY TSKF (1-313 aa encoded by BC014057)
This gene encodes a member of the short-chain dehydrogenases/reductases (SDR) family. The encoded enzyme contains a conserved catalytic domain and likely functions as an oxidoreductase. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dehydrogenase/reductase (SDR family) member 1; Dehydrogenase/reductase SDR family member 1; Protein SDR19C1; Short chain dehydrogenase/reductase family 19C member 1; short chain dehydrogenase/reductase family 19C, member 1
Gene Aliases: 1110029G07Rik; AW112170; D14Ertd484e; DHRS1; SDR19C1
UniProt ID: (Human) Q96LJ7, (Mouse) Q99L04
Entrez Gene ID: (Human) 115817, (Mouse) 52585, (Rat) 290234
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support