Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QGWFHPCAR LSVRMSSTGI DRKGVLANRV AVVTGSTSGI GFAIARRLAR DGAHVVISSR KQQNVDRAMA KLQGEGLSVA GIVCHVGKAE DREQLVAKAL EHCGGVDFLV CSAGVNPLVG STLGTSEQIW DKILSVNVKS PALLLSQLLP YMENRRGAVI LVSSIAAYNP VVALGVYNVS KTALLGLTRT LALELAPKDI RVNCVVPGII KTDFSKVFHG NESLWKNFKE HHQLQRIGES EDCAGIVSFL CSPDASYVNG ENIAVAGYST RL (1-271 aa encoded by BC007339)
DHRS2 displays NADPH-dependent dicarbonyl reductase activity in vitro with 3,4-Hexanedione, 2,3-Heptanedione and 1-Phenyl-1,2-propanedione as substrates. DHRS2 do not reductase activity is displayed in vitro with steroids, retinoids and sugars as substrates. This protein may inhibit cell replication.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dehydrogenase/reductase (SDR family) member 2; dehydrogenase/reductase member 2; Dehydrogenase/reductase SDR family member 2, mitochondrial; Dicarbonyl reductase HEP27; Protein D; Protein SDR25C1; SDR family member; Short chain dehydrogenase/reductase family 25C member 1; short chain dehydrogenase/reductase family 25C, member 1; short-chain alcohol dehydrogenase family member
Gene Aliases: 5430405K24Rik; DHRS2; HEP27; SDR25C1
UniProt ID: (Human) Q13268
Entrez Gene ID: (Human) 10202, (Mouse) 71412
Molecular Function: dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support