Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MLFWVLGLL ILCGFLWTRK GKLKIEDITD KYIFITGCDS GFGNLAARTF DKKGFHVIAA CLTESGSTAL KAETSERLRT VLLDVTDPEN VKRTAQWVKN QVGEKGLWGL INNAGVPGVL APTDWLTLED YREPIEVNLF GLISVTLNML PLVKKAQGRV INVSSVGGRL AIVGGGYTPS KYAVEGFNDS LRRDMKAFGV HVSCIEPGLF KTNLADPVKV IEKKLAIWEQ LSPDIKQQYG EGYIEKSLDK LKGNKSYVNM DLSPVVECMD HALTSLFPKT HYAAGKDAKI FWIPLSHMPA ALQDFLLLKQ KAELANPKAV (1-319 aa encoded by BC058883)
DHRS9 is a 3-alpha-hydroxysteroid dehydrogenase that converts 3-alpha-tetrahydroprogesterone (allopregnanolone) to dihydroxyprogesterone and 3-alpha-androstanediol to dihydroxyprogesterone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3-alpha hydroxysteroid dehydrogenase; 3-alpha-HSD; 3alpha-HSD; dehydrogenase/reductase (SDR family) member 9; Dehydrogenase/reductase SDR family member 9; epithelial retinol dehydrogenase; NADP-dependent retinol dehydrogenase/reductase; RDH-E2; RDH-TBE; RDHL; Retinol dehydrogenase; Retinol dehydrogenase 15; retinol dehydrogenase homolog; Short chain dehydrogenase/reductase family 9C member 4; short chain dehydrogenase/reductase family 9C, member 4; Short-chain dehydrogenase/reductase retSDR8; Tracheobronchial epithelial cell-specific retinol dehydrogenase
Gene Aliases: 3-alpha-HSD; 3ALPHA-HSD; C730025I08Rik; DHRS9; eRolDH; RDH-E2; RDH-TBE; RDH15; RDHL; RDHTBE; RETSDR8; SDR9C4; UNQ835/PRO1773
UniProt ID: (Human) Q9BPW9, (Rat) Q8VD48, (Mouse) Q58NB6
Entrez Gene ID: (Human) 10170, (Rat) 170635, (Mouse) 241452
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support