Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGKNKKHKA PAAAVVRAAV SASRAKSAEA GIAGEAQSKK PVSRPATAAA AAAGSREPRV KQGPKIYSFN STNDSSGPAN LDKSILKVVI NNKLEQRIIG VINEHKKQNN DKGMISGRLT AKKLQDLYMA LQAFSFKTKD IEDAMTNTLL YGGDLHSALD WLCLNLSDDA LPEGFSQEFE EQQPKSRPKF QSPQIQATIS PPLQPKTKTY EEDPKSKPKK EEKNMEVNMK EWILRYAEQQ NEEEKNENSK SLEEEEKFDP NERYLHLAAK LLDAKEQAAT FKLEKNKQGQ KEAQEKIRKF QREMETLEDH PVFNPAMKIS HQQNERKKPP VATEGESALN FNLFEKSAAA TEEEKDKKKE PHDVRNFDYT ARSWTGKSPK QFLIDWVRKN LPKSPNPSFE KVPVGRYWKC RVRVIKSEDD VLVVCPTILT EDGMQAQHLG ATLALYRLVK GQSVHQLLPP TYRDVWLEWS DAEKKREELN KMETNKPRDL FIAKLLNKLK QQQQQQQQHS ENKRENSEDP EESWENLVSD EDFSALSLES ANVEDLEPVR NLFRKLQSTP KYQKLLKERQ QLPVFKHRDS IVETLKRHRV VVVAGETGSG KSTQVPHFLL EDLLLNEWEA SKCNIVCTQP RRISAVSLAN RVCDELGCEN GPGGRNSLCG YQIRMESRAC ESTRLLYCTT GVLLRKLQED GLLSNVSHVI VDEVHERSVQ SDFLLIILKE ILQKRSDLHL ILMSATVDSE KFSTYFTHCP ILRISGRSYP VEVFHLEDII EETGFVLEKD SEYCQKFLEE EEEVTINVTS KAGGIKKYQE YIPVQTGAHA DLNPFYQKYS SRTQHAILYM NPHKINLDLI LELLAYLDKS PQFRNIEGAV LIFLPGLAHI QQLYDLLSND RRFYSERYKV IALHSILSTQ DQAAAFTLPP PGVRKIVLAT NIAETGITIP DVVFVIDTGR TKENKYHESS QMSSLVETFV SKASALQRQG RAGRVRDGFC FRMYTRERFE GFMDYSVPEI LRVPLEELCL HIMKCNLGSP EDFLSKALDP PQLQVISNAM NLLRKIGACE LNEPKLTPLG QHLAALPVNV KIGKMLIFGA IFGCLDPVAT LAAVMTEKSP FTTPIGRKDE ADLAKSALAM ADSDHLTIYN AYLGWKKARQ EGGYRSEITY CRRNFLNRTS LLTLEDVKQE LIKLVKAAGF SSSTTSTSWE GNRASQTLSF QEIALLKAVL VAGLYDNVGK IIYTKSVDVT EKLACIVETA QGKAQVHPSS VNRDLQTHGW LLYQEKIRYA RVYLRETTLI TPFPVLLFGG DIEVQHRERL LSIDGWIYFQ APVKIAVIFK QLRVLIDSVL RKKLENPKMS LENDKILQII TELIKTENN
DNA helicase and probable main scaffold component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair; according to PubMed:20687897 the contribution to DNA double-strand break repair appears to be largely indirect through transcriptional regulation. Recruited by YY1 to YY1-activated genes, where it acts as an essential coactivator. Binds DNA. In vitro, has double-stranded DNA-dependent ATPase activity. Involved in UV-damage excision repair, DNA replication and chromosome segregation during normal cell division cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase DHX29; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 29; DEAH (Asp-Glu-Ala-His) box polypeptide 29; DEAH box protein 29; Nucleic acid helicase DDXx
Gene Aliases: DDX29; DHX29
UniProt ID: (Human) Q63HN3
Entrez Gene ID: (Human) 54505
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support