Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This is a rabbit polyclonal anti-DHX32 antibody raised against part of the C-terminal chain of human DHX32.
Immunogen sequence: LGSSSSGKF FCLYTEEFAS KDMTPLKPAE MQEANLTSMV LFMKRIDIAG LGHCDFMNRP APESLMQALE DLDYLAALDN DGNLSEFGII MSEFPLDPQL SKSILASCEF DCVDEVLTIA AMVTAPNCFS HVPHGAEEAA LTCWKTFLHP EGDHFTLISI YKAYQDTTLN SSSEYCVEKW CRDYFLNCSA LRMADVIRAE LLEIIKRIEL PYAEPAFGSK ENTLNIKKAL LSGYFMQIAR DVDGSGNYLM LTHKQVAQLH PLSGYSITKK MPEWVLFHKF SISENNYIRI TSEISPELFM QLVPQYYFSN LPPSESKDIL QQVVDHLSPV STMNKEQQMC ETCPETEQRC TLQ (392-743 aa encoded by BC002473)
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants, but the full length nature of one of the variants has not been defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32; DEAD/H box 32; DEAD/H helicase-like protein 1; DEAD/H helicase-like protein-1; DEAH (Asp-Glu-Ala-His) box polypeptide 32; DEAH box protein 32; DHLP1; helicase DDX32; HuDDX32; MuDDX32; Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
Gene Aliases: 3110079L04Rik; 4732469F02Rik; AA408140; DDX32; DHLP1; DHX32
UniProt ID: (Human) Q7L7V1, (Mouse) Q8BZS9
Entrez Gene ID: (Human) 55760, (Mouse) 101437, (Rat) 361667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support