Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The immunogen of this antibody is the C-terminal of isoform 2 of DATF1 (DIDO1).
Immunogen sequence: ADGTDCTSI GTIEQKSSED QGIKGRIEKA ANPSGKKKLK IFQPVIEAPG ASKCIGPGCC HVAQPDSVYC SNDCILKHAA ATMKFLSSGK EQKPKPKEKM KMKPEKPSLP KCGAQAGIKI SSVHKRPAPE KKETTVKKAV VVPARSEALG KEAACESSTP SWASDHNYNA VKPEKTAAPS PSLLYKCSGK YLYSLHPSLI A (345-544 aa encoded by BC000770)
Dio-1 (Death Inducer-Obliterator-1) is a putative transcription factor that contains two zinc finger motifs. Dio-1 translocates to the nucleus, and activates apoptosis during limb development. Programmed cell death, a highly regulated form of apoptosis, plays an important role in determining the amount of tissue, the shape, and the definition of each digit during limb development. Dio-1 expression is upregulated when an apoptotic signal is detected, and subsequently apoptosis is induced. This process is similar to the expression of NFkB and NGF in response to external signals. Dio-1 expression is suppressed by caspase inhibitors and Bcl-2 expression. This supports the theory that Dio-1 functions in the onset of programmed cell death.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DATF-1; death associated transcription factor 1; death inducer-obliterator-2; death inducer-obliterator-3; Death-associated transcription factor 1; Death-inducer obliterator 1; DIO-1
Gene Aliases: 6720461J16Rik; BYE1; C20orf158; D130048F08Rik; DATF-1; DATF1; dido; DIDO1; DIDO2; DIDO3; DIO-1; DIO1; dJ885L7.8; KIAA0333
UniProt ID: (Human) Q9BTC0, (Mouse) Q8C9B9
Entrez Gene ID: (Human) 11083, (Mouse) 23856, (Rat) 362286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support