Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 6
Antibody detects endogenous levels of total DIO1.
The protein encoded by this gene is a thiol-requiring propylthiouracil-sensitive oxidoreductase. It activates thyroid hormone by converting the prohormone thyroxine (T4) by outer ring deiodination (ORD) to bioactive 3,3',5-triiodothyronine (T3). It also degrades both hormones by inner ring deiodination (IRD). Alternative splicing results in multiple transcript variants encoding different isoforms. Some, but not all, isoforms contain a selenocysteine (Sec) residue encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Additional transcript variants have been described but are not supported by experimental evidence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5DI; DIOI; hDido1; iodothyronine deiodinase type 1; thyroxine 5'-deiodinase; thyroxine deiodinase; thyroxine deiodinase type I (selenoprotein); thyroxine deiodinase, type I; Type 1 DI; Type I iodothyronine deiodinase; Type-I 5'-deiodinase
Gene Aliases: 5DI; D1; DIO1; ITDI1; TXDI1
UniProt ID: (Human) P49895, (Mouse) Q61153, (Rat) P24389
Entrez Gene ID: (Human) 1733, (Mouse) 13370, (Rat) 25430
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support