Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The exosome is involved in a multitude of cellular RNA processing and degradation events. DIS3, also known as exosome complex exonuclease RRP44, is a ribonuclease that acts directly in the processing, turnover, and surveillance of a large number of distinct RNA species. DIS3 localizes to both the cytoplasm and the nucleus and contains one PINc domain. It is widely expressed with highest expression in testis and is required for processing of 7S pre-RNA into a mature nuclear complex and, ultimately, for proper mitotic progression. Abnormal expression levels of DIS3 may be associated with colon cancer, suggesting a role for DIS3 in tumorigenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA555G22.1; DIS3 exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 mitotic control homolog; Exosome complex exonuclease RRP44; exosome component 11; mitotic control protein dis3 homolog; Protein DIS3 homolog; Ribosomal RNA-processing protein 44; RP11-342J4.3
Gene Aliases: 2810028N01Rik; DIS3; dis3p; EXOSC11; KIAA1008; RRP44
UniProt ID: (Human) Q9Y2L1, (Mouse) Q9CSH3
Entrez Gene ID: (Human) 22894, (Mouse) 72662
Molecular Function: exoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support