Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31718 has been tested in indirect ELISA.
DIS3L2 encodes a protein that is similar in sequence to 3'/5' exonucleolytic subunits of the RNA exosome. The exosome is a large multimeric ribonucleotide complex responsible for degrading various RNA substrates. Several transcript variants, some protein-coding and some not, have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DIS3 mitotic control homolog-like 2; DIS3-like exonuclease 2; DIS3L2; family with sequence similarity 6, member A; hDIS3L2
Gene Aliases: DIS3L2; FAM6A; hDIS3L2; PRLMNS
UniProt ID: (Human) Q8IYB7
Entrez Gene ID: (Human) 129563
Molecular Function:
exoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support