Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KKQERKVRVK SPRTQVGSEY RESWPPNSKL PHSQSSPTVS STCTKVLYFT DRSLTPFMVN IPKRLEEVTL KDFKAAIDRE GNHRYHFKAL D
Antigen sequence identity: Mouse 89%; Rat 88%
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil protein DIX1; Coiled-coil-DIX1; DIX domain-containing protein 1; Dixin
Gene Aliases: CCD1; DIXDC1; KIAA1735
UniProt ID: (Human) Q155Q3
Entrez Gene ID: (Human) 85458
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support