Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KLNSIKSSLG GETPGQAANR SAGMYQGLAF GGSKKGKNLG QAYPCSSDKE CEVGRYCHSP HQGSSACMVC RRKKKRCHRD GMCCPSTRCN NGICIPVTES ILTPHIPALD GTRHRDRNHG HYSNHDLGWQ NLGRPHTKMS HIKGHEGDPC
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. Activity of this protein is also modulated by binding to the Wnt co-receptor LDL-receptor related protein 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dickkopf 2; dickkopf 2 homolog; Dickkopf gene 2; dickkopf homolog 1; dickkopf homolog 2; dickkopf homolog 4; dickkopf related protein-2; Dickkopf-2; Dickkopf-related protein 2; dkk-2; hDkk-2; mDkk-2
Gene Aliases: DKK-2; DKK2; Dkk4; UNQ682/PRO1316
UniProt ID: (Human) Q9UBU2, (Mouse) Q9QYZ8
Entrez Gene ID: (Human) 27123, (Mouse) 56811, (Rat) 295445
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support