Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SKFKKLMKQG GAALEGSALA NGRALSAGSP PVPPGWNPNS SSGKGSGGNA GSYIPSYTSW YPSAHQEAMQ QPQL*
This gene encodes a member of a homeobox transcription factor gene family similiar to the Drosophila distal-less gene. The encoded protein is localized to the nucleus where it may function as a transcriptional regulator of signals from multiple TGF-beta superfamily members. The encoded protein may play a role in the control of craniofacial patterning and the differentiation and survival of inhibitory neurons in the forebrain. This gene is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 2. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DII B; distal-less homeo box 1; Homeobox protein DLX-1
Gene Aliases: Dlx; Dlx-1; DLX1
UniProt ID: (Human) P56177, (Mouse) Q64317
Entrez Gene ID: (Human) 1745, (Mouse) 13390
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support