Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MSGSFDRKLS SILTDISSSL SCHAGSKDSP TLPESSVTDL GYYSAPQHDY YSGQPYGQTV NPYTYHHQFN LNGLAGTGAY SPKSEYTYGA SYRQYGAYRE QPLPAQDPVS VKEEPEAEVR MVNGKPKKVR KPRTIYSSYQ LAALQRRFQK AQYLALPERA ELAAQLGLTQ TQVKIWFQNR RSKFKKLYKN GEVPLEHSPN NSDSMACNSP PSPALWDTSS HSTPAPARSQ LPPPLPYSAS PSYLDDPTNS WYHAQNLSGP HLQQQPPQPA TLHHASPGPP PNPGAVY
The distal-less homeobox 3 (DLX3) protein is a putative transcriptional activator that has been implicated during development and differentiation of epithelial tissues. This is a homeodomain transcription factor in vertebrates, related to Distal-less in Drosophila, which is expressed in differentiating epidermal cells, in neural crest, hair follicles, dental epithelium, mesenchyme, the optic and olfactory placodes, limb bud, placenta, and in the cement gland, which is located in the extreme anterior neural plate in Xenopus embryos.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein DLX-3
Gene Aliases: AI4; DLX3; TDO
UniProt ID: (Human) O60479
Entrez Gene ID: (Human) 1747
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support