Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: GELAERQQKL AQMLSRLQKI SEEYNVAVFV TNQMTADPGA TMTFQADPKK PIGGHILAHA STTRISLRKG RGELRIAKIY DSPEMPENEA TFAITAGGIG DAK
DMC1 is essential for meiotic homologous recombination. Genetic recombination in meiosis plays an important role in generating diversity of genetic information. The product of this protein is structurally and evolutionary related to the products of the yeast RAD51 and E. coli RecA genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: disrupted meiotic cDNA1, yeast, homolog of; DMC 1; DMC1 dosage suppressor of mck1 homolog, meiosis-specific homologous recombination; DMC1 homologue; HsLim15; Meiotic recombination protein DMC1/LIM15 homolog; MGC150472; MGC150473; RP1-199H16.4
Gene Aliases: dJ199H16.1; DMC1; DMC1H; LIM15
UniProt ID: (Human) Q14565
Entrez Gene ID: (Human) 11144, (Rat) 362960
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support