Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MREQLKGHET QTTCWDHPKM TELYQSLADL NNVRFSAYRT AMKLRRLQKA LCLDLLSLSA ACDALDQHNL KQNDQPMDIL QIINCLTTIY DRLEQEHNNL VNVPLCVDMC LNWLLNVYDT GRTGRIRVLS FKTGIISLCK AHLEDKYRYL FKQVASSTGF CDQRRLGLLL HDSIQIPRQL GEVASFGGSN IEPSVRSCFQ FANNKPEIEA ALFLDWMRLE PQSMVWLPVL HRVAAAETAK HQAKCNICKE CPIIGFRYRS LKHFNYDICQ SCFFSGRVAK GHKMHYPMVE YCTPTTSGED VRDFAKVLKN KFRTKRYFAK HPRMGYLPVQ TVLEGDNMET PVTLINFWPV DSAPASSPQL SHDDTHSRIE HYASRLAEME NSNGSYLNDS ISPNESIDDE HLLIQHYCQS LNQDSPLSQP RSPAQILISL ESEERGELER ILADLEEENR NLQAEYDRLK QQHEHKGLSP LPSPPEMMPT SPQSPRDAEL IAEAKLLRQH KGRLEARMQI LEDHNKQLES QLHRLRQLLE QPQAEAKVNG TTVSSPSTSL QRSDSSQPML LRVVGSQTSD SMGEEDLLSP PQDTSTGLEE VMEQLNNSFP SSRGHNVGSL FHMADDLGRA MESLVSVMTD EEGAE
Dystrophin is the 427kDa protein product of the DMB/BMD gene located on the X chromosome at position Xp21. Western blotting and immunohistochemistry are the two established methods for the detection of abnormalities of dystrophin expression in muscle biopsies. Dystrophin abnormalities are thought to occur in 100% of patients with DMD/BMD, although genetic abnormalities may only be detected in up to 65% of cases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Duchenne muscular dystrophy (DMD); Dystrophin; GS1-19O24.1; Muscular dystrophy Duchenne and Becker types; OTTHUMP00000215592
Gene Aliases: BMD; CMD3B; DMD; DXS142; DXS164; DXS206; DXS230; DXS239; DXS268; DXS269; DXS270; DXS272; MRX85
UniProt ID: (Human) P11532
Entrez Gene ID: (Human) 1756
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support